Eisco® 8-Piece Slotted Mass Set with Rack, Stainless Steel
Supplier: Wards
PH0037FSS
470343-864EA 111.15 USD
470343-864
Eisco® 8-Piece Slotted Mass Set with Rack, Stainless Steel
Weights Calibration Weights
Set of 8 stainless steel slotted masses with a metal and plastic rack with handle.
  • Includes 8 individually calibrated slotted masses totaling 1100 g: (1) 10 g, (2) 20 g, (1) 50 g, (1) 100 g, (2) 200 g and (1) 500 g
  • Made from high-grade stainless steel
  • Slotted for stacking on mass hangers (available separately)
  • Each piece is stamped with weight for easy identification
  • Includes sturdy metal/plastic rack with handle
This 8-piece slotted mass set is made from high-quality stainless steel and includes a storage rack with handle. Each piece is slotted for stacking on mass hangers (not included). Slotted masses included: (1) 10 g, (2) 20 g, (1) 50 g, (1) 100 g, (2) 200 g, and (1) 500 g. The convenient metal and plastic storage rack with metal carrying handle is included.