9-Piece Organic Chemistry Kit
Supplier: Wards
CH0876
470343-812EA 106.19 USD
470343-812
9-Piece Organic Chemistry Kit
Labware Sets
With a 14/23 joint size, this organic chemistry kit consists of 9 essential pieces made of borosilicate glass, providing the necessary tools for a range of experiments and ensuring durability and chemical resistance.
  • This set contains 9 items with 14/23 ground joints for maximum versatility
  • Glassware pieces are made of high quality borosilicate glass
  • Set 27 BU/M. Housed in die cut foam lined box
  • Kit consists of: 1 Flask (pear shaped, 50 ml), 1 still head, 1 liebig condenser, 1 screwcap adapter, 1 receiver adapter, 1 air leak/steam inlet tube, 1 dropping funnel 50 ml with GP rotaflow tap, 1 thermometer
Stored in die cut foam lined box. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.