Eisco Acrylic Bell Jar
Supplier: Wards
CH0145B
470343-798EA 53.65 USD
470343-798
Eisco Acrylic Bell Jar
Jars Bell Jars
An economical and safer option to traditional glass bell jars.
  • Made of high-quality acrylic
  • Lighter than traditional glass bell jars
  • No risk of shatter
  • For use in vacuum experiments
  • Ideal for student use
This acrylic bell jar can be used in many experiments including demonstrating the behavior of sound within a vacuum. Accepts a 3.5 cm (1.37") stopper, not included.