Eisco Iodine Flasks with Penny-Head Stopper
Supplier: Wards
CH0434C CH0434A CH0434B CH0434D
470343-778EA 22.15 USD
470343-778 470343-946 470345-556 470345-702
Eisco Iodine Flasks with Penny-Head Stopper
Flasks Iodine Flasks
Glass iodine flasks with ground sockets and interchangeable stopper made of borosilicate 3.3 glass.
  • Withstands high temperatures and thermal shocks
  • Ungraduated, with ground socket
  • Includes interchangeable ground glass stopper
  • Funnel-shaped cup can be filled with water with stopper inserted to prevent vapor from escaping
Iodine flasks with ground joint and interchangeable stopper used for iodine value determination. Made of durable high quality, heavy duty 3.3 borosilicate glass with the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C