Eisco Glass Reagent Bottle with Wide Neck and Stopper, 1000 ml
Supplier: Wards
CH0156FW
470343-776EA 16.3 USD
470343-776
Eisco Glass Reagent Bottle with Wide Neck and Stopper, 1000 ml
Bottles Glass Bottles
Suitable for mixing and storing samples, liquids, and reagents.
  • Wide neck for easy pouring and decanting
  • Includes a tight fitting glass stopper
  • Made of high quality transparent soda glass - built to last and withstand multiple uses
Cylindrical reagent bottle made of soda glass. Features a wide 5 cm (2") mouth for easy filling and a ground glass stopper seals the bottle to secure contents and help prevent spills and evaporation. This bottle is suitable for use in mixing and storing samples, liquids, and reagents. Size: 8.5"H×4.1"Ø.