Vacuum Distillation Set
Supplier: Wards
CH1028
470343-678EA 130.79 USD
470343-678
Vacuum Distillation Set
Distillation Components Distillation Sets
5-piece set with 24/29 joints for precise vacuum distillation, made of durable borosilicate glass.
  • Vacuum Distillation set made of high quality borosilicate glass
  • Set includes 5 pieces: 250 ml round bottom flask, still head, Liebig condenser, adaptor receiver vacuum, 100 ml Erlenmeyer flask
  • All joint sizes are 24/29
  • All pieces are made of high quality, heavy duty borosilicate glass 3.3
  • Perfect for any school or professional laboratory