Organic Chemistry and Distillation 9 Piece Set
Supplier: Wards
CH0880
470343-654EA 143.59 USD
470343-654
Organic Chemistry and Distillation 9 Piece Set
Labware Sets
Combining essential organic chemistry and distillation components, this nine-piece set offers versatility and convenience in laboratory work, all crafted from high-quality borosilicate glass for durability and optimal performance.
  • Every piece of glassware you need to do basic organic chemistry and distillation
  • Made of Borosilicate 3.3 glass
  • All glassware is jointed (14/23)
  • Made for preparations up to 30 to 40 g
Nine-piece set includes: (1) dropping funnel - 100 ml, (1) air leak/steam inlet tube, (1) Liebig condenser, (1) receiver adapter, (1) stopper, (1) flask, round bottom - 250 ml, (1) cone/screwcap adapter, (1) still head, (1) flask, pear shaped - 100 ml, (1) filter funnel - 24 mm. diameter perforated. Eisco borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.