Eisco Plastic Graduated Beaker, 10000 ml
Supplier: Wards
CH0139H
470343-642EA 48.65 USD
470343-642
Eisco Plastic Graduated Beaker, 10000 ml
Beakers
Extra-large size for a variety of applications.
  • Made of high-quality, heat and chemical-resistant polypropylene
  • Non-drip spout for easy pouring
  • Excellent optical clarity
  • Raised graduations and specifications
Extra-large 10000 ml measuring beaker. Autoclavable and microwavable with superior chemical durability. It can withstand temperatures between −20 to +135 °C. Easy-to-read blue screen-printed 1000 ml graduation lines, marked every 2000 ml.