Innovating Science® Anesthefly Kit
Supplier: Ward's Science
IS3005
470343-474EA 20.3 USD
470343-474
Innovating Science® Anesthefly Kit
Educational Classroom Kits and Activities
Learn how to anesthetize small insects without killing or sterilizing.
  • Students will anesthetize Drosophila melanogaster for 40 to 50 minutes
  • Aligned to NGSS Practices
Anesthetize Drosophila melanogaster fruit fly and other small insects for at least 40 to 50 minutes without killing or sterilizing. Includes instructions.
Delivery information: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Methanol/Methyl isobutyl ketone, which is known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.