Innovating Science® Thin Layer Chromatography
Supplier: Ward's Science
IS2504
470343-434EA 107.4 USD
470343-434
Innovating Science® Thin Layer Chromatography
Educational Classroom Kits and Activities
Learn about thin layer chromatography, an analytical tool to separate similar compounds.
  • Students will use thin layer chromatography to separate various plant leaf pigments with organic solvents.
  • Aligned to NGSS Practices
Chromatography is an analytical tool used to separate similar compounds. Thin-layer chromatography utilizes a sheet coated with silica gel. The samples are applied to the silica gel sheet and separated by migrating an appropriate solvent up the sheet.
This type of separation is called adsorption chromatography because the separation of similar compounds is based on the selective adsorption of the compounds on the silica gel solid phase.In this lab samples of various plant leaf materials may be extracted or the red and green leaf extracts included may be used. The samples of extracted leaf pigments are applied to a silica gel sheet and separated using a mixture of organic solvents.
Delivery information: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Methanol, which is known to the State of California to cause reproductive harm. For more information go to http://www.P65Warnings.ca.gov.