Innovating Science® Density and Displacement
Supplier: Ward's Science
IS2001
470343-424EA 72.7 USD
470343-424
Innovating Science® Density and Displacement
Educational Classroom Kits and Activities
Learn how to determine density and displacement.
  • Students will determine the density of four metals by determining their mass and displacement
  • Aligned to NGSS Practices
In this exercise, you will determine the density of four metals, similar in appearance. Mass will be determined using an Ohaus Scout® Pro electronic balance and volume will be calculated using Archimedes Principle of Displacement. The data obtained will then be compared to published density values. Kit contains enough materials for 15 groups.
Delivery information: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Lead and lead compounds and Nickel metallic, which are known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.