Backyard Brains® DIY Neuron Spikerbox
Supplier: BACKYARD BRAINS, INC.
BYBP0006
470323-212KT 97.9 USD
470323-212 470343-160
Backyard Brains® DIY Neuron Spikerbox
Educational Materials Biology Educational Materials Physiology Learning Activities
Build your own neuron spikerbox.
  • Shipped as individual components for your students to assemble
  • Estimated time from 1.5 (hobbyist) to 4 (novice) hours
If you want to bone up on your soldering skills and feel the pleasure that comes with building your very own bio amplifier to record neural signals, this kit is for you. The kit takes about 4 hours (complete novice) to 1.5 hours (electronics hobbyist) to build. You'll be recording spikes in no time.
Ordering information: Additional materials will be needed to assemble the kit. Please see the attached document for instructions.