Estes Rocket Science Starter Set
Supplier: Estes Industries
5302
470343-136EA 71.6 USD
470343-136
Estes Rocket Science Starter Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
The most complete model rocketry kit on the market.
  • Beginner-level build
  • Recommended engines: A8-3, B4-4, B6-4, C6-5
  • Max. altitude 335 m (1100')
  • Launch system, clinometer, and engines included
Experience the thrill of rocketry with the Crossfire™ ISX - a kit that is perfect for beginners and sure to impress everyone with its high-flying performance and sleek design! Are you ready for a high-flying adventure with the 'Rocket Science' model rocket kit? It has everything you need to launch and track your rocket! This kit is the perfect way to have fun while exploring STEM concepts. Using Estes standard model rocket engines (included with the kit), your rocket can reach impressive heights of up to 1100 ft.! Moreover, with the mini AltiTrak altitude tracker also included, you can measure and track its progress over multiple launches.
However, this kit isn't just about the excitement of launching rockets. It's also a fun and educational tool for learning about physics and engineering principles as you experiment with different engines and observe the reactions during launch. Whether you're a budding 'Rocket Scientist' or just looking for a unique way to have fun with STEM, the 'Rocket Science' model rocket kit is a fantastic choice.
Ordering information: Required but not included: (4) AA batteries.
Delivery information: Kit includes (1) crossfire ISX model rocket with parachute recovery, (1) Porta pad II, (1) electron beam launch controller, (1) mini AltiTrack, (1) B6-4 rocket engine, (1) C6-5 rocket engine, starters, and recovery wadding.