United Scientific™ Demonstration Lens Set
Supplier: United Scientific Supplies
DEM038 DEM050 DEM075
470339-424EA 14.1 USD
470339-424 470339-956 470340-118
United Scientific™ Demonstration Lens Set
Educational Materials Physics Educational Materials Optics Learning Activities
Introductory lens set for optics experiments.
  • Concave and convex selection
  • All focal lengths are 150 mm
The set contains six 38 mm lenses, one each of double convex, plano-convex,concavo-convex, double concave, plano-concave, and convexo-concave lens. Comes in a reusable cardboard storage box. Lenses in all sets have focal lengths on 150 mm.