Hooked Double-Ended Brass Weight Set of 9
Supplier: United Scientific Supplies
WHSET9
470339-964EA 146.8 USD
470339-964
Hooked Double-Ended Brass Weight Set of 9
Weights Calibration Weights
Brass hooked weights with a storage block.
  • Can be used on balance pans, as well as hooked in tandem for lever arm studies
  • Sturdy storage block included
Set of nine weights consisting of one each 10, 50, 100, 500, 1000 g, and two each 20 g and 200 g weights. Weights feature flat countersunk bottoms, with a double-ended hooked rod through the middle. They can be used on balance pans, as well as hooked in tandem for lever arm studies. Weights are made of polished brass and come in a storage block.
Please note: Due to its size, the 10 g weight does not include a countersunk bottom.