Gas Measuring Tube
Supplier: United Scientific Supplies
GTUBE50
470339-958EA 22.2 USD
470339-958
Gas Measuring Tube
Tubes Gas Sampling Tubes
Graduated borosilicate tube for measuring gas.
  • Borosilicate glass tube with 50 ml capacity
  • Closed at zero end for gas measurement
  • Autoclavable
Borosilicate glass tube features top-down graduations in durable white enamel, at 0.5 ml intervals, from 0.5 to 50 ml. Tube measures 650 mm long and 13 mm in diameter.