Support Stands with Rod, Porcelain, United Scientific Supplies
Supplier: United Scientific Supplies
SSP6X6 SSP7X13
470339-948EA 79 USD
470339-948 470339-342
Support Stands with Rod, Porcelain, United Scientific Supplies
Support Stands
Porcelain coated support stands.
  • Heavy porcelain base with glazed finish to prevent chemical seepage
  • Removable aluminum support rod
A hole for the removable support rod is conveniently located in the center of the base, and rubber feet are affixed to the base bottom to prevent sliding and/or damage to the work surface.