Digital Timer with Photogates, United Scientific Supplies
Supplier: United Scientific Supplies
DTPHG1 DTPHG1-P
470339-940EA 340.8 USD
470339-940 470339-974
Digital Timer with Photogates, United Scientific Supplies
Timers Digital Timers
Accurately measure acceleration due to gravity.
  • Four-digit timer designed for use with air tracks, free fall apparatuses, and other classroom equipment
  • Microprocessor-controlled, with eight programmed functions
  • Comes with two photogates
This four-digit timer is designed to be used with air tracks, free fall apparatuses, and other equipment. It is microprocessor-controlled and offers eight programmed functions to record counts, interruption timing, time between interruptions, period of oscillation, acceleration, acceleration due to gravity, velocity in collisions, and time scale output. Includes two photogates and activity guide.
Delivery information: Includes activity guide.