Beakers, New Style, United Scientific Supplies
Supplier: United Scientific Supplies
11110 11111 11112 11113 11114 11115 11116 11117
470340-016EA 19.5 USD
470340-016 470339-932 470339-898 470339-778 470339-042 470338-942 470339-566 470338-920
Beakers, New Style, United Scientific Supplies
Beakers
Polypropylene beakers with excellent clarity and very good chemical resistance.
  • Clear PP
  • Oversized tapered spouts
They are autoclavable and feature raised graduations on the outer surface. New Style beakers feature oversized, tapered spouts for easier pouring of solutions.