Aluminum Dissecting Trays with Wax
Supplier: United Scientific Supplies
DSPA01-W
470325-656EA 23.85 USD
470325-656 470339-910
Aluminum Dissecting Trays with Wax
Trays/Dishes/Pots Dissecting Trays
Made of aluminum and filled with a layer of paraffin wax, these dissecting pans will give you years of service with minimal maintenance.
  • Easy to Clean
  • Ideal Size for Smaller Specimens
  • Easy to Store
The paraffin holds T-pins securely without breaking down with repeated use. The wax resists absorption of most solutions allowing for a simple rinse and pour clean-up. This size of tray is ideal for smaller dissections like Clams, Squid, earthworm, locust, and most perch or smaller frogs.
Refill wax is available.