United Scientific™ Air Track
Supplier: United Scientific Supplies
ATRK01 ATRK02
470339-862EA 537.7 USD
470339-862 470339-680
United Scientific™ Air Track
Educational Materials Physics Educational Materials Mechanics Learning Activities
Investigate motion on a cushion of air.
  • Investigate speed, acceleration and force in linear motion; momentum and energy in collisions; and characteristics of harmonic oscillators
  • Constructed of lightweight, durable aluminum alloy; precision machining provides a smooth surface and high linearity
  • Includes a comprehensive accessory package for experiments in linear motion
  • Track length is 150cm, straight to better than 0.1 mm throughout
  • Air source (QAS001 or QAS001-V) and digital timer (DTPHG1) not included
Investigate speed, acceleration and force in linear motion; momentum and energy in collisions; and characteristics of harmonic oscillators. This air track is constructed of high quality aluminum alloy and has the advantages of being both lightweight and highly durable. Precision machining provides a smooth surface and high linearity. The track length is 150 cm, and it is straight to better than 0.1 mm throughout.
Ordering information: This item requires an air source (model QAS001 or QAS001-V), and a digital timer with photogates, model DTPHG1 (not included).
Delivery information: Includes activity guide and a comprehensive accessory package for experiments in linear motion.