Porcelain Evaporating Dish, Flat Form, United Scientific Supplies
Supplier: United Scientific Supplies
JEF035 JEF050 JEF080 JEF100 JEF200 JEF300
470340-056EA 27.1 USD
470340-056 470340-240 470339-020 470339-124 470339-792 470340-146
Porcelain Evaporating Dish, Flat Form, United Scientific Supplies
Trays/Dishes/Pots Evaporating Basins/Dishes
Shallow form flat bottom evaporating dish.
  • Shallow-form porcelain evaporating dishes with lip and flat bottom
  • Glazed inside and out, except for rim and part of outside bottom
  • Autoclavable
  • Withstand temperatures up to 1150 °C (2102 °F)
Pack of six shallow-form porcelain evaporating dishes with lip and flat bottom. Dishes are glazed inside and out, except for the rim and part of the outside bottom. Autoclavable and designed to withstand temperatures up to 1150 °C (2102 °F); temperature change rate should not exceed 200 °C (392 °F) per hour.