Unitary Wash Bottles with Colored Caps
Supplier: United Scientific Supplies
WBSET4
470339-772EA 23.4 USD
470339-772
Unitary Wash Bottles with Colored Caps
Bottles Wash Bottles
Wash bottle with color coded caps.
  • Set of 4 wash bottles with 500 ml capacity and built-in delivery tube
  • Made of translucent, pliable low-density polyethylene (LDPE)
  • Dispense solutions easily with a light squeeze
  • Graduated on outer surface
Set of four wash bottles with 500 ml capacity and built-in delivery tube. Made of translucent, pliable low-density polyethylene (LDPE). Polypropylene caps included: one each of red, yellow, blue, and white.