Racked Filter Micro Tips, United Scientific Supplies
Supplier: United Scientific Supplies
P11025 P11029 P11033 P11037
470339-704EA 108.6 USD
470339-704 470339-256 470338-972 470339-914
Racked Filter Micro Tips, United Scientific Supplies
Pipette Tips
Conveniently rack and sterile pipette tips.
  • Filters manufactured from 100% pure porous polyethylene
  • Tips made from ultrapure USP Class VI certified, virgin polypropylene
  • DNase, RNase and Pyrogen free
  • Case of 10 racked, sterile filter pipette tips
Each rack contains 96 graduated tips for a total of 960 tips. Designed to prevent liquids and aerosols from entering the body of the micropipette to protect against cross-contamination. Best suited for biological applications, these tips are non-cytotoxic, non-hemolytic, and have a 99.9% bacterial filtration efficiency (BFE).