Centrifuge Tubes, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
P10404 P10406 P10408
470339-698EA 169.2 USD
470339-698 470339-392 470340-130
Centrifuge Tubes, Sterile, United Scientific Supplies
Tubes Centrifuge Tubes
Leak free centrifuge tubes with conical bottoms.
  • Leak-proof, blue HDPE screw caps included
  • Tubes and caps feature specially-designed flat threads for vacuum-type closure that is leak-free at a pressure of >700 mm Hg
  • Highly smooth hydrophobic internal surfaces minimize wall effect
  • Maximum RCF is 18000 (50 ml)
  • Silk-screen graduations and frosted surface for easy writing
  • Radiation sterilized; DNase-, RNase-, Pyrogen- and latex-free
  • Autoclavable at 121 °C and 15 psi for 30 minutes.
Polypropylene tubes with conical bottoms and blue HDPE leak-proof screw caps included.