Deluxe Slotted Weight Set of 8, with Rack
Supplier: United Scientific Supplies
WHST08
470339-624EA 104.45 USD
470339-624
Deluxe Slotted Weight Set of 8, with Rack
Weights Calibration Weights
Slotted brass weights for use with hangers.
  • Set of 8 slotted brass weights
  • Durable rack also included
  • Also available as a set of 10 (WHST10) or 12 (WHST12)
Set of 8 high-quality slotted brass weights with durable rack.
Weights included: 1×500 g; 2× 200 g; 1×100 g; 1×50 g; 2×20 g; 1×10 g