Rack For Micro Tubes, PC, United Scientific Supplies
Supplier: United Scientific Supplies
P20201 P20202 P20203 P20204
470339-622EA 81 USD
470339-622 470339-730 470339-534 470339-064
Rack For Micro Tubes, PC, United Scientific Supplies
Racks Tube Racks
Stackable racks for microcentrifuge tubes.
  • Stackable white polycarbonate racks
  • Molded alphanumeric coding for quick identification of samples
  • Withstand temperatures from –135 to +135 °C (–211° to +275 °F); autoclavable
Pack of 8 stackable white polycarbonate racks, each designed to hold microcentrifuge tubes 1.5 and 2.0 ml in capacity. Each rack features 24 places in a 6×4 array. Molded alphanumeric coding allows for quick identification of samples. Can withstand temperatures from –135 to +135 °C. Autoclavable.