Support Rings with Opening, United Scientific Supplies
Supplier: United Scientific Supplies
UNOSR01 UNOSR02 UNOSR03
470338-886EA 5.85 USD
470338-886 470339-434 470339-556
Support Rings with Opening, United Scientific Supplies
Supports Flask Supports
Easily move separatory funnels wit this open ring design.
  • Open support rings
  • Ideal for use with funnels, boiling flasks, and separatory funnels
Steel rod, zinc-plated extension support rings. Rod diameter 0.27" (7 mm) and the straight rod portion of each is 7.25" (184 mm) long.