Well Plate, Clear Plastic, 96 Wells, United Scientific Supplies
Supplier: United Scientific Supplies
F1003-U
470339-414EA 35.4 USD
470339-414
Well Plate, Clear Plastic, 96 Wells, United Scientific Supplies
Microplates Deep Well Microplates
Clear polystyrene well plates.
  • Non sterile
  • Also available in 24-well design (F1003)
Each plate features 96 U-bottomed wells with alpha-numeric markings.
Packaging: Pack of 10 clear polystyrene well plates with lids; Individually wrapped in plastic bags.