Micropipette Tips, Low Retention, PP, United Scientific Supplies
Supplier: United Scientific Supplies
P10001 P10002 P10003 P10004 P10005 P10006
470339-860EA 37.8 USD
470339-860 470339-426 470339-876 470339-826 470339-758 470339-268
Micropipette Tips, Low Retention, PP, United Scientific Supplies
Pipette Tips
Reduce sample retention for more precision.
  • Highly homogenous surface is free of defects, preventing non-specific binding
  • Manufactured from ultrapure USP Class VI certified, virgin polypropylene
  • DNase, RNase and Pyrogen free
  • Natural color
Deliver 10X lower retention levels with viscous samples, as compared to standard pipette tips.
Packaging: Pack of 1000 graduated micropipette tips.