Battery Holder Set
Supplier: United Scientific Supplies
BHS001
470339-258EA 19.8 USD
470339-258
Battery Holder Set
Battery Holders
Build a battery powered power supply with four voltage settings.
  • Battery holder fits four D cell batteries
  • Useful for virtually any experiment requiring a low voltage power supply
  • Firmly mounted on a heavy duty base; ships fully assembled
  • Batteries not included
This battery holder fits four D cell batteries (not included) and is useful for virtually any experiment requiring a low voltage power supply. Five (5) color-coded binding posts on the base make for easy, safe, and reliable connections to hook-up cords providing outlets for 1.5, 3, 4.5 or 6 V supply voltages. Battery holder is firmly mounted on a heavy-duty base.