Microscope Slide Tray, Polystyrene
Supplier: United Scientific Supplies
62301
470339-214EA 65 USD
470339-214
Microscope Slide Tray, Polystyrene
Slide Holders
Polystyrene microscope slide tray with unique step design.
  • Holds 20 slides
  • Unique stepped cut-out design
  • Polystyrene construction
Microscope slide storage tray designed for 20 each 1×3" slides. A unique stepped cut-out design simplifies removal of slides and is designed to avoid direct contact of prepared slides with the tray surface. Polystyrene trays nest within each other. White color provides good contrast for viewing dyes and stains.
Tray dimensions: 11.25" W×7.5" D×0.75" H