Molecular Model Set, Student
Supplier: United Scientific Supplies
58012
470339-194EA 59.9 USD
470339-194
Molecular Model Set, Student
Models Molecular Biology Models
Suitable for constructing complex organic and inorganic molecules.
  • Allows student to make a wide range of molecues
  • Periodic table included
Suitable for constructing complex organic and inorganic molecules. The student set has 48 atom centers and 35 bonds. Includes a durable plastic storage box with a copy of the periodic table for easy reference.
Contents: Carbon (Black): 6, Hydrogen (White): 14, Oxygen (Red): 6, Nitrogen (Blue): 3, Sulphur (Yellow): 3, Halogen (Green): 5, Phosphorus (Purple): 1, Divalent Metals (Silver): 5, Trivalent Metals (Silver): 4, Bonds (Large): 10, Bonds (Small): 25.
Kit contains small parts, not for children under 3 years.