Test Tube Drying Rack, United Scientific Supplies
Supplier: United Scientific Supplies
77851
470339-148PK 48.35 USD
470339-148
Test Tube Drying Rack, United Scientific Supplies
Racks Tube Racks
Drying racks for tubes and plates.
  • Autoclavable
  • Inverted tubes design to minimize collection of airborne contaminants
These polypropylene racks can conveniently hold inverted tubes for drying and minimize collection or airborne contaminants. Racks can also be used to hold and dry electrophoresis and chromatography plates. Autoclavable. The 96 place rack features an 8×12 array of pegs that hold test tubes up to 13 mm in diameter.