Racked Micro Tips, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
P10111 P10113 P10117 P10148
470339-346EA 88.3 USD
470339-346 470339-866 470339-134 470339-428
Racked Micro Tips, Sterile, United Scientific Supplies
Pipette Tips
Convenient rack and sterile micropipette tips.
  • Inner radial rings enable airtight grip on tip cone
  • Manufactured in a precision molding facility from 100% virgin polypropylene
  • DNase, RNase and Pyrogen free
Case of 10 racked, sterile pipette tips. Each rack contains 96 tips, size 2 to 200 μl, for a total of 960 tips.
The highly smooth, hydrophobic surface provides low liquid retention, increased accuracy and repeatability.
Color: Yellow