UniStore™ Reagent Bottles, Narrow Mouth, PP
Supplier: United Scientific Supplies
33253 33254 33301 33302 33303 33304 33305 33253-BULK 33254-BULK 33301-BULK 33302-BULK 33303-BULK 33304-BULK 33305-BULK
76263-130PK 7.64 USD
76263-130 76263-132 76263-134 76263-136 76263-138 76263-140 76263-142 470338-912 470340-042 470339-084 470340-074 470339-926 470338-968 470338-966
UniStore™ Reagent Bottles, Narrow Mouth, PP
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Rigid
  • Autoclavable
UniStore™ narrow mouth PP reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.