United Quiet Air Source with Variable Output
Supplier: United Scientific Supplies
QAS001-V
470339-012EA 334.9 USD
470339-012
United Quiet Air Source with Variable Output
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Low noise air source for air tracks.
  • Low noise (<58 dB) air source - quieter than normal conversation
  • Ideal accessory for air tracks, air tables, and other pneumatic equipment in physics labs
This low noise (<58 dB) air source is an ideal accessory for air tracks, air tables, and other pneumatic equipment in physics labs. The 250 W motor produces clean air output with a flow rate of 35 m³/hour at a 0.70 kPa overpressure.