Culture Tubes, Disposable, with Screw-Cap Finish, United Scientific Supplies
Supplier: United Scientific Supplies
DCTSC-13100 DCTSC-16100 DCTSC-16125 DCTSC-16150 DCTSC-20125 DCTSC-20150
470339-054EA 296.9 USD
470339-054 470339-752 470339-986 470339-248 470338-998 470339-718
Culture Tubes, Disposable, with Screw-Cap Finish, United Scientific Supplies
Tubes Culture Tubes
Disposable borosilicate glass culture tube with screw cap finish.
  • Type I, Class B 5.1 expansion borosilicate glass
  • Heat-treated and securely packed to avoid contamination and increase strength
  • Chevron-shaped marking spot for easy orientation reference
Manufactured from 5.1 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements, these threaded culture tubes are suitable for a variety of lab settings. Each tube is heat-treated at temperatures exceeding 500° C and immediately packed in a shrink-wrapped cardboard tray, 250 tubes per tray. This process avoids contamination and increases the strength and durability of the glass.
Dimensional tolerances are strictly controlled for a uniform shape, including rounded bottoms and polished tops with smooth edges. Each tube has a chevron-shaped white enamel marking spot. Threaded necks accept a phenolic screw cap, not included. Available in six sizes, from 8 to 33 ml.
Please note: The capacity given below indicates the brim capacity (overflow capacity).