Slotted Weight Set of 10, with Rack
Supplier: United Scientific Supplies
WHST10
470338-918EA 92.7 USD
470338-918
Slotted Weight Set of 10, with Rack
Weights Calibration Weights
Slotted brass weights for use with hangers.
  • Set of 10 slotted brass weights
  • Durable rack also included
  • Also available as a set of 8 (WHST08) or 12 (WHST12)
Set of 10 high-quality slotted brass weights with durable rack.
Weights included: 1×500 g; 5×100 g; 1×50 g; 2×20 g; 1×10 g