United Scientific™ Inertia Demonstrator
Supplier: United Scientific Supplies
IDEM01
470338-884EA 27.05 USD
470338-884
United Scientific™ Inertia Demonstrator
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Vividly demonstrate Newton's concept of inertia.
  • Durable inertia demonstrator for classroom use
  • Demonstrator allows for 2 distinct experiments
  • Includes activity guide
This durable inertia demonstrator works reliably every time and is a great improvement on traditional versions of this classic experiment. A steel ball rests on a thin metal card on top of a pillar, and a spring-loaded catch is mounted next to the pillar. When the catch is released, a piston strikes the edge of the metal card sharply.
The card flies out from under the ball, which remains in place on top of the pillar due to its large inertial mass. The card is tethered to the aluminum base for easy recovery after the experiment. In a second experiment the cord is pulled slowly and steadily. In this case, the ball and card fall off the pillar together. Approximate product dimensions: 5×3×2".