ReptiFeast Live Redworms (Eisenia fetida)
Supplier: SEGREST FARMS
572-0073-3
470338-846EA 10.7 USD
470338-846
ReptiFeast Live Redworms (Eisenia fetida)
Specimens Live Specimens Animals - Living Material
Red wigglers make a good live food to feed Axolotls and other aquatic amphibians and reptiles.
  • Chemical-free; safe for feeding to pets
  • Great Source of Protein
  • Also make great composting worms
ReptiFeast® live food was designed by animal keepers for animal keepers. The grab-n-go cup will make it quick and easy easy to keep alive, just store the cup refrigerated at 40°F.
Delivery information: These products contain living materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.