Estes Regular Engine Mounts
Supplier: Estes Industries
3158
470338-820CS 241.09 USD
470338-820
Estes Regular Engine Mounts
Educational Classroom Kits and Activities
Mounting kit for Estes standard engines and rocket body tubes.
  • Fits standard-size 19 mm diameter engines
  • Converts to BT-50, BT-55, BT-56, and BT-60 body tubes
  • Easy to assemble, with complete instructions included
This engine mount kit fits BT-50, 55, 56, 60 body tubes. They can also be used to make a conversion mount for lightweight D powered rockets.