Fluorescent Slime Using Polyvinyl Alcohol
Supplier: Ward's Science
IS7041
470338-590EA 24.95 USD
470338-590
Fluorescent Slime Using Polyvinyl Alcohol
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Create your own fluorescent slime while understanding polymer chemistry.
  • Understand and demonstrate polymer chemistry through a synthesis reaction
  • Aligned to NGSS practices
  • Cross cutting concepts: structure and function, patterns
  • Disciplinary core ideas: PS1.A, PS1.B
  • Learn engineering practices: analyzing and interpreting data, obtaining, evaluating, and communicating information
Create your own fluorescent polymers (Slime) in this great classroom demonstration. Mixture is pink under regular light and glows orange under a black light. Included pigment will make this polymer activity easy to see and memorable for your students!