Oxidation of Glycerin
Supplier: Ward's Science
IS7028
470338-582EA 25 USD
470338-582
Oxidation of Glycerin
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
A small amount of potassium permanganate is placed in a petri dish. A few drops of glycerin are added and after a few seconds, a puff of smoke and violet flames are produced.
  • Understand and demonstrate an oxidation reaction
  • Aligned to NGSS practices
Perform an oxidation reaction with glycerin and potassium permanganate and witness the violet flame it produces.