War of the Acids
Supplier: Ward's Science
IS7044
470338-574EA 66 USD
470338-574
War of the Acids
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Understand strong versus weak acids, conjugate bases, and the effect of different acids on pH.
  • Understand and demonstrate acid-base chemistry
  • Aligned to NGSS practices
Many people think that all acids are the same. Often times when students hear the phrases strong acid and weak acid they will confuse these concepts with high/low concentration of the acid. This demonstration kit has been designed to visualize the differences between strong and weak acids. Activities include demonstrations of differences in pH values, effect of conjugate base and vigor of reactions.
Caution: WARNING: This product can expose you to chemicals including methanol/methyl isobutyl ketone, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov