Negative Coefficient of Solubility
Supplier: Ward's Science
IS7016
470338-564EA 24 USD
470338-564
Negative Coefficient of Solubility
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Explain how there are exceptions to the rules of chemistry through a negative coefficient of solubility.
  • Understand and demonstrate negative coefficients of solubility
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns
  • Disciplinary core ideas: PS1.A, PS1.B
  • Learn engineering practices: analyzing and interpreting data
Explain how there are exceptions to the rules of chemistry. Heat a solution and watch it form a precipitate. Watch it go back into solution as it cools.