Solid Fuel
Supplier: Ward's Science
IS7025
470338-560EA 28.5 USD
470338-560
Solid Fuel
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Two solutions are poured together in a beaker and form a gel. The gel can be ignited with a match. Commercially this gel is sold as Sterno®.
  • Understand and demonstrate a solid fuel synthesis reaction
  • Aligned to NGSS practices
  • Disciplinary core ideas: PS1.A, PS1.B
Caution: WARNING: This product can expose you to chemicals including methanol, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov