Kitronik Discovery Kit
Supplier: NEWARK ELECTRONICS
5666
470338-528EA 23.25 USD
470338-528
Kitronik Discovery Kit
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Take your first steps in physical computing with the Kitronik Discovery Kit for micro:bit.
  • Simple build: no soldering or tools required!
  • Five fun experiments with detailed tutorial book
  • Simple, block-based coding
  • Extend learning with challenge experiments, or design your own project
The Kitronik Discovery kit for BBC microbit is a great way to get started with both programming and electronics. The Kit contains five experiments that start very simply, building up to simulating real world systems as confidence grows.
The included booklet assumes no prior knowledge and contains detailed information about everything the new user will need to know. It covers everything from using a prototyping board to how to use the Microsoft MakeCode Editor, and everything in between. This is the ideal kit for someone who is new to the micro:bit, electronics, and coding.
The kit contains five experiments and all of the components required to complete them. Each experiment has; a complete code walk-through, a circuit diagram and a top-down breadboard view, full explanations of what is happening, and how the electronics work.
Ordering information: Required but not included: micro:bit controller.
Delivery information: Kit includes (1) breadboard breakout for the BBC micro:bit, (1) Small prototype breadboard, (2) red 10 mm LEDs, (2) yellow 10 mm LEDs, (2) green 10 mm LEDs, (10) 330 Ω resistors, (1) piezo element buzzer, and (10) male to male jumper wires.