Ward's® Luria Broth with Ampicillin
Supplier: VWR International
470337-968
470337-968CS
327.8
USD
470337-968
Ward's® Luria Broth with Ampicillin
Media for Microbiology
Dehydrated Media for Microbiology
For recombinant strains of Escherichia coli that are Ampicillin resistant.
- Selects for ampicillin resistance
- Sterile, no autoclaving
- Conveniently prepared for you consistenly every time
For genetic and molecular studies using recombinant strains of Escherichia coli containing plasmids with the resistance gene to Ampicillin.
Delivery information: Prepared media will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Caution: For educational use only.