Wards® Forming Red, White, and Blue Chemical Demonstration Kit
Supplier: Ward's Science
IS7036
470337-952EA 35 USD
470337-952
Wards® Forming Red, White, and Blue Chemical Demonstration Kit
Educational Materials Chemistry Educational Materials Ionic Properties Learning Activities
Discover the different chemical reaction types via this colorful demonstration.
  • Contains enough materials for 7 demonstrations
Introduce chemical reaction types. This is a patriotic demonstration that illustrates complex ion formation, double replacement while introducing chemical reaction types.
Delivery information: Includes 1×30 ml Iron (III) chloride, 1.0 M solution; 1×140 ml Silver nitrate, 0.1 M solution; 1×140 ml Potassium ferrocyanide, 0.1 M solution; 1×140 ml Potassium thiocyanate, 0.002 M solution.